Rhodamine 6G perchlorate 99%, pure laser grade

Warning

AC41903-5000
200032-748EA 49.1 USD
200032-748
Rhodamine 6G perchlorate 99%, pure laser grade
Rhodamine 6G perchlorate
Formula: C₂₈H₃₁ClN₂O₇
Melting Pt: 263…265 °C
Storage Temperature: Ambient
MDL Number: MFCD00013117
CAS Number: 13161-28-9
EINECS: 236-099-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR