4-(2-Pyridylazo)resorcinol 97+%

Warning

Synonyms: 4-(2'-Pyridylazo)resorcinol

AC41859-0250 AC41859-0050 AC41859-0010
200023-466EA 29.9 USD
200023-466 103514-878 200010-092
4-(2-Pyridylazo)resorcinol 97+%
4-(2-Pyridylazo)resorcinol
Formula: C₁₁H₉N₃O₂
MW: 215.21
Melting Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00044237
EINECS: 214-528-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR