(R)-(+)-Pulegone 92% pract.

Synonyms: p-Menth-4(8)-en-3-one(R)-p-Menth-4(8)-en-3-one(R)-5-Methyl-2-(propan-2-ylidene)cyclohexanone(R)-2-Isopropylidene-5-methylcyclohexanone(+)-Pulegone

AC41850-1000 AC41850-5000
103466-410EA 449 USD
103466-410 200014-278
(R)-(+)-Pulegone 92% pract.
(R)-(+)-Pulegone
Formula: C₁₀H₁₆O
MW: 152.23 g/mol
Boiling Pt: 97 °C (12 mmHg)
Density: 0.931 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00063000
CAS Number: 89-82-7
EINECS: 201-943-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR