2-(Propyloxy)ethanol 98%, pure

Danger

Synonyms: Propyl glycol2-PropoxyethanolEthylene glycol monopropyl ether

AC41840-0050 AC41840-0010
102776-512EA 202 USD
102776-512 200003-958
2-(Propyloxy)ethanol 98%, pure
2-(Propyloxy)ethanol
Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Melting Pt: –70 °C
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00072645
CAS Number: 2807-30-9
EINECS: 220-548-6
UN: 1986
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR