Potassium bis(trimethylsilyl)amide 15% (w/w) 0.7 M in toluene, AcroSeal™

Danger

AC41823-1000
103528-642EA 70.2 USD
103528-642
Potassium bis(trimethylsilyl)amide 15% (w/w) 0.7 M in toluene, AcroSeal™
Potassium bis(trimethylsilyl)amide
Formula: C₆H₁₈KNSi₂
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00010330
CAS Number: 40949-94-8
EINECS: 424-100-2
UN: 2920
ADR: 8,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR