Phloxin B ≥85%, pure, certified

Synonyms: Sodium 2',4',5',7'-tetrabromo-4,5,6,7-tetrachloro-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)CyanosinePhloxineAcid Red 92Eosine Blue2′,4′,5′,7′-Tetrabromo-4,5,6,7-tetrachlorofluoresceinEosin S extra bluishCI 45410

AC41787-0250
200003-944EA 111 USD
200003-944
Phloxin B ≥85%, pure, certified
Phloxin B
Formula: C₂₀H₂Br₄Cl₄Na₂O₅
MW: 829.6344 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070627
CAS Number: 18472-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR