N,N,N',N',N''-Pentamethyldiethylenetriamine 98+%

Danger

Synonyms: 1,1,4,7,7-PentamethyldiethylenetriamineN1-(2-(Dimethylamino)ethyl)-N1,N2,N2-trimethylethane-1,2-diamine

AC41695-1000 AC41695-5000 AC41695-0250
102776-164EA 175 USD
102776-164 200015-464 200008-366
N,N,N',N',N''-Pentamethyldiethylenetriamine 98+%
N,N,N',N',N''-Pentamethyldiethylenetriamine
Formula: C₉H₂₃N₃
MW: 173.30 g/mol
Melting Pt: 18 °C
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00014876
CAS Number: 3030-47-5
EINECS: 221-201-1
UN: 2922
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR