Sodium palmitate 98%

Synonyms: Palmitic Acid Sodium Salt

AC41670-0050 AC41670-0250 AC41670-1000
103518-888EA 689 USD
103518-888 200013-870 200032-434
Sodium palmitate 98%
Sodium palmitate
Formula: C₁₆H₃₂O₂
MW: 278.41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002749
CAS Number: 408-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR