4-Phenylazophenol 97%

Warning

Synonyms: 4-(Phenyldiazenyl)phenol

AC41736-0250
200026-596EA 45.9 USD
200026-596
4-Phenylazophenol 97%
4-Phenylazophenol
Formula: C₁₂H₁₀N₂O
MW: 198.23 g/mol
Melting Pt: 150…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00002330
CAS Number: 1689-82-3
EINECS: 216-880-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR