Naphthol AS-BI phosphate 93%

Warning

Synonyms: Naphthol AS-BI Phosphoric acid solution6-Bromo-3-((2-methoxyphenyl)carbamoyl)naphthalen-2-yl dihydrogen phosphate7-Bromo-3-hydroxy-2-naphthoic-o-anisidide phosphate

AC41531-1000 AC41531-0010
200020-000EA 197 USD
200020-000 103514-826
Naphthol AS-BI phosphate 93%
Naphthol AS-BI phosphate
Formula: C₁₈H₁₅BrNO₆P
MW: 452.1926 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00004058
CAS Number: 1919-91-1
EINECS: 217-645-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR