1-Naphthalenethiol ≥99%

Supplier: ACROS ORGANICS MS

Synonyms: 1-Thionaphthol

AC41527-0050
200008-734EA 212.73 USD
200008-734
1-Naphthalenethiol ≥99%
1-Naphthalenethiol
Formula: C₁₀H₈S
MW: 160.24 g/mol
Density: 1,15 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00039599
CAS Number: 529-36-2

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6790 to 1.6810 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR