(Methyl)triphenoxyphosphonium iodide 95%
Synonyms:
Methyltriphenoxyphosphonium iodide
AC41502-0050
AC41502-0500
200019-802EA
39.1
USD
200019-802
103514-822
(Methyl)triphenoxyphosphonium iodide 95%
(Methyl)triphenoxyphosphonium iodide
Formula:
C₁₉H₁₈IO₃P Melting Pt: 141…146 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00011911 CAS Number: 17579-99-6 EINECS: 241-551-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...