Methyl green zinc chloride salt, pure, certified

Warning

Synonyms: CI 42590Methyl green zinc chloride double saltMethyl GreenEthyl Green

AC41438-0250
200019-728EA 101 USD
200019-728
Methyl green zinc chloride salt, pure, certified
Methyl Green zinc chloride salt
Formula: C₂₇H₃₅BrClN₃·xZnCl₂
MW: 653.24
Melting Pt: 233 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00151094
CAS Number: 7114-03-6
EINECS: 230-415-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR