Methyl-α-chloroacrylate 98+% stabilized

Danger


Stabilization: Stabilised with hydroquinone (HQ)

AC41416-0250
200013-270EA 288 USD
200013-270
Methyl-α-chloroacrylate 98+% stabilized
Methyl-alpha-chloroacrylate

Appearance: Clear colorless to yellow Liquid

Formula: C₄H₅ClO₂
Boiling Pt: 52 °C (50 mmHg)
Density: 1.189 g/cm³
Flash Pt: 33 °C
Storage Temperature: Refrigerator
CAS Number: 80-63-7
UN: 3489
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR