2-Methylbutyryl chloride 98%

Danger

Synonyms: DL-2-Methylbutyryl chloride

AC41415-0250 AC41415-0010 AC41415-1000 AC41415-0050
102774-938EA 38.9 USD
102774-938 200018-900 102773-826 200043-370
2-Methylbutyryl chloride 98%
2-Methylbutyryl chloride
Formula: C₅H₉ClO
Flash Pt: 20 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041726
CAS Number: 57526-28-0
EINECS: 260-787-3
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR