Isobutyl methacrylate 99% stabilized

Warning

Synonyms: Methacrylic acid Isobutyl ester

AC41268-0025 AC41268-0010
102776-022EA 162 USD
102776-022 200019-386
Isobutyl methacrylate 99% stabilized
Isobutyl methacrylate
Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Melting Pt: –37 °C
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00008931
CAS Number: 97-86-9
EINECS: 202-613-0
UN: 2283
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR