Imidazole-4,5-dicarboxylic acid 99%

Warning

Synonyms: 4,5-Imidazoledicarboxylic acid1H-Imidazole-4,5-dicarboxylic acid

AC41225-0250
200003-844EA 138 USD
200003-844
Imidazole-4,5-dicarboxylic acid 99%
Imidazole-4,5-dicarboxylic acid
Formula: C₅H₄N₂O₄
MW: 156.10 g/mol
Melting Pt: 265 °C
Storage Temperature: Ambient
MDL Number: MFCD00005200
CAS Number: 570-22-9
EINECS: 209-327-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR