4-Hydrazinobenzoic acid 98%

Warning

Synonyms: 4-Hydrazinylbenzoic acid

AC41179-0050 AC41179-0250 AC41179-5000 AC41179-1000
200020-396EA 95 USD
200020-396 103512-466 200038-520 103512-464
4-Hydrazinobenzoic acid 98%
4-Hydrazinobenzoic acid
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007581
CAS Number: 619-67-0
EINECS: 210-609-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR