Heparin sodium salt

Synonyms: Reviparin sodium

AC41121-0010
200003-828EA 230.54 USD
200003-828
Heparin sodium salt
Heparin sodium salt

Appearance: White to almost white Powder

Storage Temperature: Ambient
MDL Number: MFCD00081689
CAS Number: 9041-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR