Guanosine 99%

Danger

Synonyms: 2-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one

AC41113-1000 AC41113-5000 AC41113-0250
103530-306EA 542 USD
103530-306 200003-824 103533-106
Guanosine 99%
Guanosine
Formula: C₁₀H₁₃N₅O₅
MW: 283.24 g/mol
Melting Pt: 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010182
CAS Number: 118-00-3
EINECS: 204-227-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR