(S)-(-)-2,3-Epoxy-1-propanol 97%, 98% ee

Danger

Synonyms: (S)-(-)-Glycidol(S)-Oxiran-2-ylmethanol

AC41100-0250 AC41100-0010 AC41100-0050
102777-286EA 347 USD
102777-286 102775-482 102773-944
(S)-(-)-2,3-Epoxy-1-propanol 97%, 98% ee
(S)-(-)-2,3-Epoxy-1-propanol
Formula: C₃H₆O₂
MW: 74.08 g/mol
Melting Pt: –54 °C
Flash Pt: 66 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00074874
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR