Geraniol 99%

Danger

Synonyms: (E)-3,7-Dimethylocta-2,6-dien-1-ol

AC41090-1000 AC41090-5000 AC41090-0025 AC41090-0250 AC41090-0050
103512-436EA 294 USD
103512-436 200012-570 103512-434 200043-602 200012-568
Geraniol 99%
Geraniol
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 229…230 °C (757 mmHg)
Melting Pt: –15 °C
Density: 0.878 g/cm³
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00002917
CAS Number: 106-24-1
EINECS: 203-377-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR