Ethylene glycol dimethacrylate (ethylene dimethacrylate) 98% stabilized

Warning

Synonyms: Ethylene dimethacrylateEthane-1,2-diyl bis(2-methylacrylate)

AC40992-2500 AC40992-0010 AC40992-0051
200003-802EA 76.3 USD
200003-802 103514-810 103514-812
Ethylene glycol dimethacrylate (ethylene dimethacrylate) 98% stabilized
Ethylene glycol dimethacrylate (Ethylene dimethacrylate)
Formula: C₁₀H₁₄O₄
MW: 198.21 g/mol
Boiling Pt: 98…100 °C (5 mmHg)
Melting Pt: –40 °C
Density: 1.05 g/cm³
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008590
CAS Number: 97-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR