5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulfonic acid monosodium salt hydrate 98+%, pure

Warning

Synonyms: 3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4',4''-disulphonic acid monosodium salt hydrate3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4',4''-disulfonic acid monosodium salt hydratePDT-sulfonate monosodium salt hydratePDT disulfonate monosodium salt hydratePDT-sulphonate monosodium salt hydrateFerroZine iron reagent hydratePDT disulphonate monosodium salt hydrate

AC41057-0010 AC41057-0050 AC41057-0250
103464-180EA 661 USD
103464-180 200003-810 103464-178
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulfonic acid monosodium salt hydrate 98+%, pure
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulphonic acid monosodium salt hydrate

FerroZine™ iron reagent

Formula: C₂₀H₁₃N₄NaO₆S₂·H₂O
MW: 492.47 g/mol (anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150794
CAS Number: 69898-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR