trans,trans-Dibenzylideneacetone 98%

Synonyms: (1E,4E)-1,5-Diphenylpenta-1,4-dien-3-one

AC40900-0050 AC40900-0250 AC40900-1000
103514-324EA 29.5 USD
103514-324 200033-818 200033-820
trans,trans-Dibenzylideneacetone 98%
trans,trans-Dibenzylideneacetone
Formula: C₁₇H₁₄O
MW: 234.30 g/mol
Melting Pt: 107…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00004790
CAS Number: 35225-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR