1,3-Diphenoxybenzene 99%
AC40884-0250
AC40884-0050
200063-934EA
50
USD
200063-934
200037-858
1,3-Diphenoxybenzene 99%
1,3-Diphenoxybenzene
Formula:
C₁₈H₁₄O₂ MW: 262.31 g/mol Melting Pt: 58…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003036 CAS Number: 3379-38-2 EINECS: 222-181-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...