Dinonyl phthalate mixture of isomers 96%

AC40877-0250 AC40877-1000
103514-314EA 381 USD
103514-314 200006-348
Dinonyl phthalate mixture of isomers 96%
Dinonyl phthalate
Formula: C₂₆H₄₂O₄
Boiling Pt: 279…287 °C (760 mmHg)
Density: 0.97 g/cm³
Flash Pt: 216 °C
Storage Temperature: Ambient
MDL Number: MFCD00036237
CAS Number: 84-76-4
EINECS: 201-560-0
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR