Lithium tetrachlorocuprate 0.1 M in THF
|
Danger
|
Synonyms:
Dilithium tetrachlorocuprate
AC40783-1000
AC40783-5000
AC40783-0250
200057-438EA
36.82
USD
200057-438
200057-436
103533-094
Lithium tetrachlorocuprate 0.1 M in THF
Lithium tetrachlorocuprate
Formula:
Cl₄CuLi₂ MW: 219.24 g/mol Density: 0.91 g/cm³ Flash Pt: –17 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011081 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...