Digitonin

Danger

AC40756-5000 AC40756-0050
200003-778EA 238 USD
200003-778 103528-576
Digitonin
Digitonin
Formula: C₅₆H₉₂O₂₉
Melting Pt: 240 °C
Storage Temperature: Ambient
EINECS: 234-255-6
UN: 3462
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR