Diethyl terephthalate 98%

Synonyms: Diethyl benzene-1,4-dicarboxylate

AC40745-0250 AC40745-1000
200025-014EA 85.4 USD
200025-014 200052-554
Diethyl terephthalate 98%
Diethyl terephthalate
Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
Melting Pt: 42…45 °C
Flash Pt: 117 °C
Storage Temperature: Ambient
MDL Number: MFCD00039891
CAS Number: 636-09-9
EINECS: 211-249-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR