Diethyl oxalacetate sodium salt 95% pract.

Warning

Synonyms: Sodium diethyl oxalacetateSodium Diethyl OxalacetateOxalacetic Acid Diethyl Ester Sodium Salt

AC40737-5000 AC40737-1000
103518-858EA 142 USD
103518-858 200012-050
Diethyl oxalacetate sodium salt 95% pract.
Diethyl oxalacetate sodium salt
Formula: C₈H₁₂O₅
MW: 210.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00035571
CAS Number: 40876-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR