Diethyl chlorophosphate 95%

Danger

AC40724-0250 AC40724-1000 AC40724-5000
102773-918EA 36.8 USD
102773-918 200003-776 103528-574
Diethyl chlorophosphate 95%
Diethyl chlorophosphate
Formula: C₄H₁₀ClO₃P
Boiling Pt: 81 °C (6 mmHg)
Density: 1.19 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00009075
CAS Number: 814-49-3
EINECS: 212-396-4
UN: 2927
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR