Dicyclohexylphenylphosphine 95%

Warning

Synonyms: Dicyclohexyl(phenyl)phosphine

AC40708-0010
200043-182EA 70.1 USD
200043-182
Dicyclohexylphenylphosphine 95%
Dicyclohexylphenylphosphine
Formula: C₁₈H₂₇P
MW: 274.38 g/mol
Melting Pt: 54…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00003854
CAS Number: 6476-37-5
EINECS: 229-334-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR