Di-tert-butyl malonate 98%

AC40662-0500 AC40662-0100
200006-370EA 60.3 USD
200006-370 103514-780
Di-tert-butyl malonate 98%
Di-tert-butyl malonate
Formula: C₁₁H₂₀O₄
Boiling Pt: 110…111 °C (22 mmHg)
Melting Pt: –7 °C
Density: 0.966 g/cm³
Flash Pt: 88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008810
CAS Number: 541-16-2
EINECS: 208-769-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR