Diacetin 50% tech. remainder triacetin and monoacetin

Synonyms: Glycerol alpha,alpha'-Diacetate2-Hydroxypropane-1,3-diyl diacetate

AC40630-5000 AC40630-0050
200043-126EA 15.7 USD
200043-126 200018-716
Diacetin 50% tech. remainder triacetin and monoacetin
Diacetin

Appearance: Clear colorless Liquid

Formula: C₇H₁₂O₅
MW: 176.17 g/mol
Boiling Pt: 280 °C (760 mmHg)
Density: 1.1875 g/cm³
Flash Pt: 141 °C
Storage Temperature: Ambient
MDL Number: MFCD00008717
CAS Number: 25395-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR