Decyltrimethylammonium bromide 99%

Warning

Synonyms: (1-Decyl)trimethylammonium bromideN,N,N-Trimethyldecan-1-aminium bromide

AC40623-0250 AC40623-1000
200018-700EA 266 USD
200018-700 200011-944
Decyltrimethylammonium bromide 99%
Decyltrimethylammonium bromide
Formula: C₁₃H₃₀BrN
MW: 280.30 g/mol
Melting Pt: 243 °C
Storage Temperature: Ambient
MDL Number: MFCD00041973
EINECS: 218-219-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR