Coumarin 343, pure laser grade

Synonyms: 2,3,6,7-Tetrahydro-11-oxo-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizine-10-carboxylic Acid

AC40568-5000 AC40568-1000
102774-232EA 44.8 USD
102774-232 200006-116
Coumarin 343, pure laser grade
Coumarin 343

Appearance: Orange Crystalline powder

Formula: C₁₆H₁₅NO₄
MW: 285.30 g/mol
Melting Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00051335
CAS Number: 55804-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR