7-(Ethylamino)-4,6-dimethylcoumarin 99% laser grade

Warning

Synonyms: 7-(Ethylamino)-4,6-dimethyl-2H-chromen-2-oneCoumarin 450Coumarin 2

AC40548-5000
200006-114EA 62.7 USD
200006-114
7-(Ethylamino)-4,6-dimethylcoumarin 99% laser grade
7-(Ethylamino)-4,6-dimethylcoumarin
Formula: C₁₃H₁₅NO₂
MW: 217.2637 g/mol
Melting Pt: 168…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00006860
EINECS: 247-446-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR