tert-Butyl isocyanide 97%

Danger

Synonyms: t-Butylisocyanide

AC40360-0050 AC40360-0010
103528-528EA 146 USD
103528-528 200003-730
tert-Butyl isocyanide 97%
tert-Butyl isocyanide
Formula: C₅H₉N
MW: 83.13 g/mol
Boiling Pt: 91 °C
Density: 0.73 g/cm³
Flash Pt: –2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000002
CAS Number: 7188-38-7
EINECS: 230-553-5
UN: 1992
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR