2-Butanone oxime 99%

Danger

AC40333-0010 AC40333-0250
103533-080EA 99.9 USD
103533-080 102773-764
2-Butanone oxime 99%
2-Butanone oxime
Formula: C₄H₉NO
Boiling Pt: 72 °C (25 mmHg)
Melting Pt: –30 °C
Density: 0.923 g/cm³
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00013935
CAS Number: 96-29-7
EINECS: 202-496-6
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR