Desyl bromide 97%

Danger

Synonyms: 2-Bromo-2-phenylacetophenone

AC40317-0250 AC40317-0050
200028-546EA 253 USD
200028-546 200016-376
Desyl bromide 97%
Desyl bromide
Formula: C₁₄H₁₁BrO
Melting Pt: 53…58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000136
CAS Number: 1484-50-0
EINECS: 216-057-1
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Appearance White to beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 53 to 58 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR