Bromohydroquinone 94% pract.

Warning

Synonyms: 2-Bromobenzene-1,4-diol

AC40304-0010
200042-786EA 27.6 USD
200042-786
Bromohydroquinone 94% pract.
Bromohydroquinone
Formula: C₆H₅BrO₂
MW: 189.01 g/mol
Melting Pt: 104…110 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00041747
CAS Number: 583-69-7
EINECS: 209-516-2

Order Now

Specification Test Results

Appearance Beige to brown Crystalline powder
Infrared spectrum Conforms
Melting point 104 to 110 °C
Assay 39.7 to 42.7 % (bromine, by combustion)
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR