Betaine monohydrate ≥99% for analysis

Supplier: ACROS ORGANICS MS

AC40219-5000 AC40219-1000
103514-714EA 150.71 USD
103514-714 200003-720
Betaine monohydrate ≥99% for analysis
Betaine monohydrate
Formula: C₅H₁₁NO₂·1H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150010
CAS Number: 590-47-6
EINECS: 203-490-6

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying 11 % to 15 % (140°C, 3 h)
Sulfated ash ≤0.1 %
Solubility (10% in H2O) Clear colorless solution
Titration with HClO4 99.0 to 101.0 % (On dry substance)
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤200 ppm
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR