(S)-(-)-4-Benzyl-2-oxazolidinone 99%, >99% ee

Warning

Synonyms: (S)-4-Benzyloxazolidin-2-one(S)-4-Benzyl-2-oxazolidinone

AC40216-0250 AC40216-0025 AC40216-1000
200053-456EA 236 USD
200053-456 103514-712 200021-414
(S)-(-)-4-Benzyl-2-oxazolidinone 99%, >99% ee
(S)-(-)-4-Benzyl-2-oxazolidinone
Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Melting Pt: 87…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00064496
CAS Number: 90719-32-7

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 87 to 90 °C
HPLC ≥98.5 %
Enantiomeric excess ≥99 %
Specific optical rotation -60° to -64° (20 °C, 589 nm) (c=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR