1-(2-Aminoethyl)piperazine 99%
|
Danger
|
AC40076-1000
AC40076-0025
AC40076-5000
102776-614EA
230
USD
102776-614
200008-304
200039-828
1-(2-Aminoethyl)piperazine 99%
1-(2-Aminoethyl)piperazine
Formula:
C₆H₁₅N₃ Melting Pt: –19 °C Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005971 CAS Number: 140-31-8 EINECS: 205-411-0 UN: 2815 ADR: 8,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4990 to 1.5010 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...