N-(2-Aminoethyl)morpholine 98+%

Danger

Synonyms: 2-Morpholinoethanamine4-(2-Aminoethyl)morpholine

AC40075-1000 AC40075-0050 AC40075-0250
200010-090EA 177 USD
200010-090 102774-806 200005-318
N-(2-Aminoethyl)morpholine 98+%
N-(2-Aminoethyl)morpholine
Formula: C₆H₁₄N₂O
MW: 130.19 g/mol
Melting Pt: 24 °C
Flash Pt: 175 °C
Storage Temperature: Ambient
MDL Number: MFCD00006182
CAS Number: 2038-03-1
EINECS: 218-011-6
UN: 2923
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to white to yellow Liquid or solid
Infrared spectrum Conforms
Refractive index 1.4740 to 1.4780 (20 °C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR