Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Aminobenzenesulfonic acid ≥95%

Supplier: ACROS ORGANICS MS

Danger
Synonyms: Aniline-2-sulphonic acidOrthanilic acid

AC40064-0250 AC40064-5000 AC40064-1000
200007-220EA 25.54 USD InStock
200007-220 102776-448 102774-568
2-Aminobenzenesulfonic acid ≥95%
2-Aminobenzenesulfonic acid


Formula: C₆H₇NO₃S
MW: 173.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007705
CAS Number: 88-21-1
EINECS: 201-810-9
UN: 2585
ADR: 8,III
Order Now
Specification Test Results
Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥94 %


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty