Alizarin red S (sodium salt), pure, certified

Synonyms: Sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonateSodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulphonateAlizarin sulfonic acid disodium saltC.I. 58005Alizarin sulphonic acid disodium salt

AC40048-0250
200003-692EA 66.5 USD
200003-692
Alizarin red S (sodium salt), pure, certified
Alizarin Red S (sodium salt)
Formula: C₁₄H₇NaO₇S
MW: 342.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013049
CAS Number: 130-22-3
EINECS: 204-981-8

Order Now

Specification Test Results

Appearance Yellow to orange to brown Powder
Infrared spectrum Conforms
UV Lambda max 554 to 558 nm
UV Lambda max 590 to 599 nm
Visual transition interval from pH 3.5 (yellow) to pH 6.5 (pink)
Visual transition interval from pH 9.4 (orange-brown) to pH 12.0 (violet)
BSC Certification Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR