9-Acridone 99%

Warning

Synonyms: Acridin-9(10H)-one9(10H)-Acridone

AC40025-0250 AC40025-0100
200030-228EA 380 USD
200030-228 200017-938
9-Acridone 99%
9-Acridone
Formula: C₁₃H₉NO
MW: 195.22 g/mol
Melting Pt: 360…362 °C
Storage Temperature: Ambient
MDL Number: MFCD00005019
CAS Number: 578-95-0
EINECS: 209-434-7

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Titration with TBAH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR