4-Fluoro-o-toluonitrile 97%

Warning

AC39993-0010 AC39993-0050
102775-824EA 100.4 USD
102775-824 102774-514
4-Fluoro-o-toluonitrile 97%
4-Fluoro-o-toluonitrile
Formula: C₈H₆FN
MW: 135.14 g/mol
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD03095106
CAS Number: 147754-12-9
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 68 to 72 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR